Assay Details
Gene Symbol: NUDT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927480.4 | XP_001927515.2 | 531 | 129 | ||
GenBank mRNA | AK234900.1 | - | 2 - 3 | 497 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
