Assay Details
Gene Symbol: RFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121967.5 | XP_003122015.3 | 427 | 103 | ||
GenBank mRNA | AK235403.1 | - | 1 - 2 | 334 | 103 | |
AK395340.1 | - | 1 - 2 | 334 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
