Assay Details
Gene Symbol: RFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121967.5 | XP_003122015.3 | 676 | 170 | ||
GenBank mRNA | AK235403.1 | - | 3 - 4 | 583 | 170 | |
AK395340.1 | - | 3 - 4 | 583 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
