Assay Details
Gene Symbol: HACD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121884.6 | XP_003121932.2 | 172 | 73 | ||
XM_021063085.1 | XP_020918744.1 | 161 | 73 | |||
GenBank mRNA | AK235492.1 | - | 1 - 2 | 176 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
