Assay Details
Gene Symbol: RCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927777.6 | XP_001927812.4 | 5604 | 84 | ||
GenBank mRNA | AK235578.1 | - | 1218 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
