Assay Details
Gene Symbol: EMR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021083944.1 | XP_020939603.1 | 1195 | 67 | ||
XM_021083945.1 | XP_020939604.1 | 1146 | 67 | |||
XM_021083946.1 | XP_020939605.1 | 1196 | 67 | |||
XM_021083947.1 | XP_020939606.1 | 1196 | 67 | |||
XM_021083948.1 | XP_020939607.1 | 1196 | 67 | |||
XM_021083949.1 | XP_020939608.1 | 1196 | 67 | |||
XM_021083950.1 | XP_020939609.1 | 1196 | 67 | |||
GenBank mRNA | AK230928.1 | - | 10 - 11 | 1120 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
