Assay Details
Gene Symbol: SMUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927219.5 | XP_001927254.1 | 101 | 72 | ||
GenBank mRNA | AK235795.1 | - | 1 - 2 | 31 | 72 | |
AK389170.1 | - | 1 - 2 | 134 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001927219.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
