Assay Details
Gene Symbol: FOSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122519.3 | XP_003122567.1 | 97 | |||
GenBank mRNA | AK236246.1 | - | 2 - 3 | 541 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
