Assay Details
Gene Symbol: PYCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021090543.1 | XP_020946202.1 | 202 | 90 | ||
GenBank mRNA | AK236710.1 | - | 1 - 2 | 127 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
