Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094215.1 | XP_020949874.1 | 1819 | 107 | ||
XM_021094216.1 | XP_020949875.1 | 1670 | 107 | |||
GenBank mRNA | AK237049.1 | - | 6 - 7 | 799 | 107 | |
AK237671.1 | - | 4 - 5 | 544 | 107 | ||
AK345450.1 | - | 3 - 4 | 378 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
