Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094215.1 | XP_020949874.1 | 2171 | 101 | ||
XM_021094216.1 | XP_020949875.1 | 2022 | 101 | |||
XM_021094217.1 | XP_020949876.1 | 2037 | 101 | |||
GenBank mRNA | AK237049.1 | - | 8 - 9 | 1151 | 101 | |
AK237671.1 | - | 6 - 7 | 896 | 101 | ||
AK345450.1 | - | 5 - 6 | 730 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
