Assay Details
Gene Symbol: NFKB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021072741.1 | XP_020928400.1 | 2004 | 86 | ||
XM_021072742.1 | XP_020928401.1 | 2087 | 86 | |||
XM_021072743.1 | XP_020928402.1 | 2004 | 86 | |||
XM_021072744.1 | XP_020928403.1 | 2001 | 86 | |||
XM_021072745.1 | XP_020928404.1 | 2107 | 86 | |||
XM_021072746.1 | XP_020928405.1 | 2001 | 86 | |||
GenBank mRNA | AK237079.1 | - | 3 - 4 | 439 | 86 | |
AK351644.1 | - | 8 - 9 | 801 | 86 | ||
AK396815.1 | - | 15 - 16 | 2066 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
