Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078142.1 | XP_020933801.1 | 577 | 61 | ||
XM_021078144.1 | XP_020933803.1 | 357 | 61 | |||
GenBank mRNA | AK237411.1 | - | 1 - 2 | 261 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021078142.1, XM_021078144.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
