Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078142.1 | XP_020933801.1 | 1044 | 103 | ||
XM_021078143.1 | XP_020933802.1 | 543 | 103 | |||
XM_021078144.1 | XP_020933803.1 | 821 | 103 | |||
XM_021078145.1 | XP_020933804.1 | 564 | 103 | |||
GenBank mRNA | AK237411.1 | - | 6 - 7 | 728 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
