Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078142.1 | XP_020933801.1 | 1531 | 108 | ||
XM_021078143.1 | XP_020933802.1 | 1030 | 108 | |||
XM_021078144.1 | XP_020933803.1 | 1308 | 108 | |||
XM_021078145.1 | XP_020933804.1 | 1051 | 108 | |||
GenBank mRNA | AK237411.1 | - | 8 - 9 | 1549 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
