Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087371.1 | XP_020943030.1 | 888 | 70 | ||
GenBank mRNA | AK237423.1 | - | 6 - 7 | 694 | 70 | |
AK240445.1 | - | 6 - 7 | 694 | 70 | ||
AK240464.1 | - | 6 - 7 | 723 | 70 | ||
AK351101.1 | - | 6 - 7 | 697 | 70 | ||
AK393030.1 | - | 6 - 7 | 696 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
