Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021076030.1 | XP_020931689.1 | 616 | 71 | ||
GenBank mRNA | AK237629.1 | - | 2 - 3 | 566 | 71 | |
AK238208.1 | - | 2 - 3 | 544 | 71 | ||
AK238458.1 | - | 2 - 3 | 566 | 71 | ||
AK393146.1 | - | 2 - 3 | 578 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
