Assay Details
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021083559.1 | XP_020939218.1 | 807 | 61 | ||
XM_021083560.1 | XP_020939219.1 | 728 | 61 | |||
GenBank mRNA | AK237632.1 | - | 1 - 2 | 586 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
