Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096391.1 | XP_020952050.1 | 7595 | 63 | ||
XM_021096401.1 | XP_020952060.1 | 7535 | 63 | |||
XM_021096411.1 | XP_020952070.1 | 7505 | 63 | |||
XM_021096419.1 | XP_020952078.1 | 7583 | 63 | |||
XM_021096425.1 | XP_020952084.1 | 7562 | 63 | |||
XM_021096427.1 | XP_020952086.1 | 7541 | 63 | |||
XM_021096435.1 | XP_020952094.1 | 7481 | 63 | |||
XM_021096441.1 | XP_020952100.1 | 7322 | 63 | |||
XM_021096453.1 | XP_020952112.1 | 6205 | 63 | |||
GenBank mRNA | AK237731.1 | - | 1 - 2 | 151 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
