Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005659906.3 | XP_005659963.1 | 2929 | 102 | ||
XM_005659907.3 | XP_005659964.1 | 2309 | 102 | |||
XM_005659909.3 | XP_005659966.1 | 3010 | 102 | |||
XM_013993367.2 | XP_013848821.1 | 2772 | 102 | |||
XM_021073967.1 | XP_020929626.1 | 4287 | 102 | |||
XM_021073969.1 | XP_020929628.1 | 4272 | 102 | |||
XM_021073974.1 | XP_020929633.1 | 4158 | 102 | |||
XM_021073975.1 | XP_020929634.1 | 4131 | 102 | |||
XM_021074004.1 | XP_020929663.1 | 2354 | 102 | |||
XM_021074012.1 | XP_020929671.1 | 2442 | 102 | |||
XM_021074016.1 | XP_020929675.1 | 2319 | 102 | |||
XM_021074027.1 | XP_020929686.1 | 2319 | 102 | |||
XM_021074030.1 | XP_020929689.1 | 2191 | 102 | |||
GenBank mRNA | AK237997.1 | - | 12 - 13 | 1287 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
