Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481554.4 | XP_003481602.1 | 396 | 90 | ||
GenBank mRNA | AK238002.1 | - | 1 - 2 | 119 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003481554.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
