Assay Details
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003133090.4 | XP_003133138.1 | 126 | 79 | ||
GenBank mRNA | AK238311.1 | - | 1 - 2 | 133 | 79 | |
AK345710.1 | - | 1 - 2 | 98 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003133090.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
