Assay Details
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003133090.4 | XP_003133138.1 | 188 | 103 | ||
GenBank mRNA | AK238311.1 | - | 2 - 3 | 195 | 103 | |
AK345710.1 | - | 2 - 3 | 160 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003133090.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
