Assay Details
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003133090.4 | XP_003133138.1 | 516 | 101 | ||
XM_021073006.1 | XP_020928665.1 | 441 | 101 | |||
GenBank mRNA | AK238311.1 | - | 4 - 5 | 523 | 101 | |
AK345710.1 | - | 4 - 5 | 488 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
