Assay Details
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481140.4 | XP_003481188.1 | 356 | 83 | ||
GenBank mRNA | AK238485.1 | - | 3 - 4 | 321 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
