Assay Details
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666209.3 | XP_005666266.2 | 353 | 71 | ||
XM_013978129.2 | XP_013833583.2 | 353 | 71 | |||
GenBank mRNA | AK231127.1 | - | 3 - 4 | 338 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
