Assay Details
Gene Symbol: RPL18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123511.5 | XP_003123559.4 | 301 | 134 | ||
XM_021083533.1 | XP_020939192.1 | 2533 | 134 | |||
XM_021083534.1 | XP_020939193.1 | 2568 | 134 | |||
GenBank mRNA | AK231188.1 | - | 2 - 3 | 232 | 134 | |
AK234580.1 | - | 2 - 3 | 236 | 134 | ||
AK237419.1 | - | 2 - 3 | 236 | 134 | ||
AK394532.1 | - | 2 - 3 | 237 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
