Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003132133.3 | XP_003132181.1 | 382 | 99 | ||
GenBank mRNA | AK231412.1 | - | 2 - 3 | 190 | 99 | |
AK399848.1 | - | 2 - 3 | 395 | 99 | ||
AY610265.1 | - | 2 - 3 | 229 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
