Assay Details
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021065066.1 | XP_020920725.1 | 1806 | 82 | ||
GenBank mRNA | AK238519.1 | - | 4 - 5 | 368 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
