Assay Details
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021065066.1 | XP_020920725.1 | 2354 | 96 | ||
GenBank mRNA | AK238519.1 | - | 9 - 10 | 916 | 96 | |
AK351265.1 | - | 9 - 10 | 916 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
