Assay Details
Gene Symbol: MCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125978.4 | XP_003126026.1 | 793 | 61 | ||
GenBank mRNA | AK238521.1 | - | 3 - 4 | 769 | 61 | |
AK396077.1 | - | 3 - 4 | 798 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
