Assay Details
Gene Symbol: MAD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129223.4 | XP_003129271.1 | 552 | 88 | ||
GenBank mRNA | AK238626.1 | - | 3 - 4 | 446 | 88 | |
AK239379.1 | - | 3 - 4 | 482 | 88 | ||
AK343950.1 | - | 3 - 4 | 426 | 88 | ||
AK347389.1 | - | 3 - 4 | 432 | 88 | ||
AK348344.1 | - | 3 - 4 | 432 | 88 | ||
AK351765.1 | - | 3 - 4 | 446 | 88 | ||
AY610160.1 | - | 3 - 4 | 437 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
