Assay Details
Gene Symbol: CLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122716.3 | XP_003122764.1 | 154 | 80 | ||
GenBank mRNA | AK238734.1 | - | 1 - 2 | 113 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003122716.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
