Assay Details
Gene Symbol: TNFSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003480815.3 | XP_003480863.1 | 418 | 55 | ||
GenBank mRNA | AK238931.1 | - | 1 - 2 | 306 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
