Assay Details
Gene Symbol: GET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123324.4 | XP_003123372.1 | 643 | 118 | ||
GenBank mRNA | AK238998.1 | - | 3 - 4 | 479 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
