Assay Details
Gene Symbol: STARD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124844.6 | XP_003124892.3 | 772 | 66 | ||
GenBank mRNA | AK239282.1 | - | 1 - 2 | 166 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
