Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064185.1 | XP_020919844.1 | 221 | 74 | ||
GenBank mRNA | AK239358.1 | - | 1 - 2 | 221 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
