Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067551.1 | XP_020923210.1 | 866 | 104 | ||
GenBank mRNA | AK239624.1 | - | 5 - 6 | 810 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
