Assay Details
Gene Symbol: CDC42EP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094975.1 | XP_020950634.1 | 699 | 76 | ||
GenBank mRNA | AK240320.1 | - | 2 - 3 | 381 | 76 | |
AK240502.1 | - | 2 - 3 | 379 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021094975.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
