Assay Details
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021079234.1 | XP_020934893.1 | 325 | 80 | ||
GenBank mRNA | AK240507.1 | - | 2 - 3 | 323 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021079234.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
