Assay Details
Gene Symbol: ATPSCKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021076853.1 | XP_020932512.1 | 459 | 97 | ||
GenBank mRNA | AK343235.1 | - | 3 - 4 | 476 | 97 | |
AK348171.1 | - | 3 - 4 | 461 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
