Assay Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129079.4 | XP_003129127.1 | 603 | 96 | ||
XM_005666732.3 | XP_005666789.1 | 603 | 96 | |||
XM_013978764.2 | XP_013834218.1 | 603 | 96 | |||
XM_021100598.1 | XP_020956257.1 | 603 | 96 | |||
XM_021100599.1 | XP_020956258.1 | 603 | 96 | |||
XM_021100601.1 | XP_020956260.1 | 603 | 96 | |||
XM_021100602.1 | XP_020956261.1 | 510 | 96 | |||
GenBank mRNA | AK343468.1 | - | 2 - 3 | 511 | 96 | |
AK350619.1 | - | 2 - 3 | 475 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021100602.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
