Assay Details
Gene Symbol: RPS27L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003480338.3 | XP_003480386.1 | 539 | 99 | ||
GenBank mRNA | AK344097.1 | - | 3 - 4 | 274 | 99 | |
AY610398.1 | - | 3 - 4 | 287 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
