Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069628.1 | XP_020925287.1 | 1942 | 84 | ||
GenBank mRNA | AK344324.1 | - | 1 - 2 | 106 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021069628.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
