Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069626.1 | XP_020925285.1 | 121 | |||
XM_021069628.1 | XP_020925287.1 | 121 | ||||
GenBank mRNA | AK344324.1 | - | 2 - 3 | 300 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021069626.1, XM_021069628.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
