Assay Details
Gene Symbol: CH25H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671263.3 | XP_005671320.1 | 1584 | 109 | ||
XM_021073088.1 | XP_020928747.1 | 2135 | 109 | |||
XM_021073089.1 | XP_020928748.1 | 3797 | 109 | |||
GenBank mRNA | AK344355.1 | - | 1 | 724 | 109 | |
AK390552.1 | - | 1 | 718 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
