Assay Details
Gene Symbol: SH2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087296.1 | XP_020942955.1 | 266 | 63 | ||
GenBank mRNA | AK344888.1 | - | 5 - 6 | 291 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
