Assay Details
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003359010.2 | XP_003359058.1 | 244 | 60 | ||
GenBank mRNA | AK345302.1 | - | 1 - 2 | 132 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003359010.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
