Assay Details
Gene Symbol: HHLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005670241.3 | XP_005670298.1 | 295 | 94 | ||
XM_005670242.3 | XP_005670299.1 | 459 | 94 | |||
XM_005670245.3 | XP_005670302.1 | 389 | 94 | |||
XM_005670246.3 | XP_005670303.1 | 480 | 94 | |||
XM_013982469.2 | XP_013837923.1 | 541 | 94 | |||
XM_013982470.2 | XP_013837924.1 | 295 | 94 | |||
XM_013982473.2 | XP_013837927.1 | 295 | 94 | |||
XM_021070436.1 | XP_020926095.1 | 560 | 94 | |||
XM_021070437.1 | XP_020926096.1 | 177 | 94 | |||
XM_021070438.1 | XP_020926097.1 | 412 | 94 | |||
XM_021070439.1 | XP_020926098.1 | 389 | 94 | |||
XM_021070440.1 | XP_020926099.1 | 459 | 94 | |||
XM_021070441.1 | XP_020926100.1 | 421 | 94 | |||
XM_021070452.1 | XP_020926111.1 | 295 | 94 | |||
GenBank mRNA | AK345908.1 | - | 1 - 2 | 79 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
