Assay Details
Gene Symbol: TRAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081624.1 | XP_020937283.1 | 212 | 93 | ||
XM_021081625.1 | XP_020937284.1 | 212 | 93 | |||
GenBank mRNA | AK346671.1 | - | 73 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021081624.1, XM_021081625.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
